Product Details

SNP ID
rs104894427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75004850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGCCTTCCTCAAAGAGGCTGCC[C/T]GAAAGAGGAAATTCCATGTCATTGT
Phenotype
MIM: 606454 MIM: 604395
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF2B2 PubMed Links

Gene Details

Gene
EIF2B2
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014239.3 629 Nonsense Mutation CGA,TGA R183* NP_055054.1
Gene
MLH3
Gene Name
mutL homolog 3
There are no transcripts associated with this gene.

View Full Product Details