Product Details
- SNP ID
-
rs7150025
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:96281951 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGGATGCTTTTTGTATCTCTGATA[G/T]GCAGGCACTATAATGGGGGGAAATA
- Phenotype
-
MIM: 616226
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ATG2B
PubMed Links
Gene Details
- Gene
- ATG2B
- Gene Name
- autophagy related 2B
View Full Product Details