Product Details

SNP ID
rs6823504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:435942 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAGATAATGAGATGCTGATGAGT[A/T]ATCTTATGTCACCAGGAAGTAGTTA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCA11P PubMed Links

Gene Details

Gene
ABCA11P
Gene Name
ATP binding cassette subfamily A member 11, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF721
Gene Name
zinc finger protein 721
There are no transcripts associated with this gene.

View Full Product Details