Product Details

SNP ID
rs7681990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:425850 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCCAGCGCCTGCTCTTTACAGAG[C/T]TCTAAGAACACCTGCTCCAAGGTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCA11P PubMed Links

Gene Details

Gene
ABCA11P
Gene Name
ATP binding cassette subfamily A member 11, pseudogene
There are no transcripts associated with this gene.

View Full Product Details