Product Details

SNP ID
rs6965212
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:139341719 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTCGGGGACCAAAGGACCAGCGG[G/T]GGGGGGCGCAGAGTCGCTCTGGGGA
Phenotype
MIM: 613056
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
C7orf55-LUC7L2 PubMed Links
Additional Information
For this assay, SNP(s) [rs116523410] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C7orf55-LUC7L2
Gene Name
C7orf55-LUC7L2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244584.2 Intron NP_001231513.1
Gene
FMC1
Gene Name
formation of mitochondrial complex V assembly factor 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_197964.4 Intron NP_932068.2
Gene
LUC7L2
Gene Name
LUC7 like 2, pre-mRNA splicing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244585.1 Intron NP_001231514.1
NM_001270643.1 Intron NP_001257572.1
NM_016019.4 Intron NP_057103.2

View Full Product Details