Product Details
- SNP ID
-
hCV29080672
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:138076758 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCAATTTGTAATAATGAAAGGAC[A/T]ACTATTTACTGAATAGGTATTGAGC
- Phenotype
-
MIM: 604741
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AKR1D1
PubMed Links
Gene Details
- Gene
- AKR1D1
- Gene Name
- aldo-keto reductase family 1, member D1
There are no transcripts associated with this gene.
View Full Product Details