Product Details

SNP ID
rs7195536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11348171 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAGTTAAGTTTTATTGGAATACA[C/T]TGGTAATTTACAGAATGTCTCGGGC
Phenotype
MIM: 612426
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105371083 PubMed Links

Gene Details

Gene
LOC105371083
Gene Name
uncharacterized LOC105371083
There are no transcripts associated with this gene.

Gene
RMI2
Gene Name
RecQ mediated genome instability 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152308.2 Intron NP_689521.1

View Full Product Details