Product Details

SNP ID
rs7012758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81694113 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTCTATGCTTGATGACTATTCCA[A/G]TTGATACTGGCACAAGTATGAAAAG
Phenotype
MIM: 602064
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IMPA1 PubMed Links

Gene Details

Gene
IMPA1
Gene Name
inositol monophosphatase 1
There are no transcripts associated with this gene.

Gene
SLC10A5
Gene Name
solute carrier family 10 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010893.2 860 Missense Mutation ACT,ATT T287I NP_001010893.1
Gene
ZFAND1
Gene Name
zinc finger AN1-type containing 1
There are no transcripts associated with this gene.

View Full Product Details