Product Details

SNP ID
rs7807417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:3309849 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTTTATTTTTGCCTTGTAGGCAT[A/G]TTCCCTTTTTCTAAGCGTGCTGTCT
Phenotype
MIM: 607216
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDK1 PubMed Links

Gene Details

Gene
SDK1
Gene Name
sidekick cell adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079653.1 Intron NP_001073121.1
NM_152744.3 Intron NP_689957.3
XM_011515188.1 Intron XP_011513490.1
XM_011515189.1 Intron XP_011513491.1
XM_011515190.1 Intron XP_011513492.1
XM_017011837.1 Intron XP_016867326.1
XM_017011838.1 Intron XP_016867327.1
XM_017011839.1 Intron XP_016867328.1
XM_017011840.1 Intron XP_016867329.1

View Full Product Details