Product Details

SNP ID
rs6656956
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:161766778 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTGGGACCGCCAAAAAGGCTACA[A/G]GCTATTTCCTGAGTGTGAAATGTGG
Phenotype
MIM: 605537 MIM: 604835
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATF6 PubMed Links

Gene Details

Gene
ATF6
Gene Name
activating transcription factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007348.3 Intron NP_031374.2
XM_006711224.1 Intron XP_006711287.1
XM_011509308.1 Intron XP_011507610.1
XM_011509309.1 Intron XP_011507611.1
XM_011509310.2 Intron XP_011507612.1
Gene
DUSP12
Gene Name
dual specificity phosphatase 12
There are no transcripts associated with this gene.

View Full Product Details