Product Details

SNP ID
rs7205335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:82632585 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAAATGTCATCACGCACCAGCAG[C/T]CAAGGGACTCTGAAACCCTGGGATC
Phenotype
MIM: 601364
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH13 PubMed Links

Gene Details

Gene
CDH13
Gene Name
cadherin 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220488.1 Intron NP_001207417.1
NM_001220489.1 Intron NP_001207418.1
NM_001220490.1 Intron NP_001207419.1
NM_001220491.1 Intron NP_001207420.1
NM_001220492.1 Intron NP_001207421.1
NM_001257.4 Intron NP_001248.1
XM_011522804.2 Intron XP_011521106.1
XM_017022848.1 Intron XP_016878337.1
XM_017022849.1 Intron XP_016878338.1

View Full Product Details