Product Details
- SNP ID
-
rs8048990
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:80542737 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCTATAGTTATCAAGAAAATCCTA[C/G]AATCCTGGAATACTTTGTTTTTATA
- Phenotype
-
MIM: 607168
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DYNLRB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75718465] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DYNLRB2
- Gene Name
- dynein light chain roadblock-type 2
- Gene
- LOC102724084
- Gene Name
- uncharacterized LOC102724084
There are no transcripts associated with this gene.
View Full Product Details