Product Details

SNP ID
rs7186611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:80542233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCCATTTGTGGTAGTGACATAC[A/G]ATTTCCTCTTAAAATGCATTTATTT
Phenotype
MIM: 607168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DYNLRB2 PubMed Links

Gene Details

Gene
DYNLRB2
Gene Name
dynein light chain roadblock-type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305017.1 Intron NP_001291946.1
NM_130897.2 Intron NP_570967.1
XM_006721295.3 Intron XP_006721358.1
XM_006721296.3 Intron XP_006721359.1
XM_011523370.2 Intron XP_011521672.1
Gene
LOC102724084
Gene Name
uncharacterized LOC102724084
There are no transcripts associated with this gene.

View Full Product Details