Product Details

SNP ID
rs6889575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:77186515 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGTTAGGCCTGCAACCTTTTCCC[A/G]TTGACCTGCCTGGGAAGATGACTGC
Phenotype
MIM: 603390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDE8B PubMed Links

Gene Details

Gene
PDE8B
Gene Name
phosphodiesterase 8B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029851.2 Intron NP_001025022.1
NM_001029852.2 Intron NP_001025023.1
NM_001029853.2 Intron NP_001025024.1
NM_001029854.2 Intron NP_001025025.1
NM_003719.3 Intron NP_003710.1
XM_005248621.4 Intron XP_005248678.1
XM_005248623.4 Intron XP_005248680.1
XM_005248624.4 Intron XP_005248681.1
XM_006714725.3 Intron XP_006714788.1
XM_006714726.3 Intron XP_006714789.1
XM_011543699.2 Intron XP_011542001.1
XM_011543700.2 Intron XP_011542002.1
XM_011543701.2 Intron XP_011542003.1
XM_011543702.2 Intron XP_011542004.1
XM_011543703.2 Intron XP_011542005.1
XM_011543704.2 Intron XP_011542006.1
XM_017010003.1 Intron XP_016865492.1
XM_017010004.1 Intron XP_016865493.1
XM_017010005.1 Intron XP_016865494.1
XM_017010006.1 Intron XP_016865495.1
XM_017010007.1 Intron XP_016865496.1
XM_017010008.1 Intron XP_016865497.1

View Full Product Details