Product Details

SNP ID
rs4952051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:31173320 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAAGCTACGCTTTCAAAGTGCTC[C/T]TTTCTTCTGGTACAGTACCCATCTG
Phenotype
MIM: 610229
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAPN14 PubMed Links

Gene Details

Gene
CAPN14
Gene Name
calpain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145122.1 3660 UTR 3 NP_001138594.1
NM_001321270.1 3660 UTR 3 NP_001308199.1
XM_011532863.2 3660 UTR 3 XP_011531165.1
XM_011532864.2 3660 UTR 3 XP_011531166.1
XM_011532865.1 3660 UTR 3 XP_011531167.1
XM_011532866.2 3660 UTR 3 XP_011531168.1

View Full Product Details