Product Details
- SNP ID
-
rs7605824
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:280819 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTCTATTAATGAGCTTCAGAACGC[A/G]GCAAATGGATTTGCTCTGAGGTGAG
- Phenotype
-
MIM: 171500
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201664542] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACP1
- Gene Name
- acid phosphatase 1, soluble
There are no transcripts associated with this gene.
- Gene
- FAM150B
- Gene Name
- family with sequence similarity 150 member B
- Gene
- LOC101927262
- Gene Name
- melanoma-associated antigen C1-like
There are no transcripts associated with this gene.
View Full Product Details