Product Details

SNP ID
rs7396514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1470841 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTCCCTGATGGACACTTGGGGCC[A/G]TCTCCCGGGCCAGGACGGAGGTGGC
Phenotype
MIM: 609236 MIM: 611969
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRSK2 PubMed Links

Gene Details

Gene
BRSK2
Gene Name
BR serine/threonine kinase 2
There are no transcripts associated with this gene.

Gene
MOB2
Gene Name
MOB kinase activator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172223.2 Intron NP_001165694.1
NM_053005.5 Intron NP_443731.2

View Full Product Details