Product Details

SNP ID
rs7693347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:98886923 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATGTTTCACTGTACTATAAACAG[A/G]AGTCATATGACAGAACAAAAATAAT
Phenotype
MIM: 133440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF4E PubMed Links

Gene Details

Gene
EIF4E
Gene Name
eukaryotic translation initiation factor 4E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130678.1 Intron NP_001124150.1
NM_001130679.1 Intron NP_001124151.1
NM_001968.3 Intron NP_001959.1
XM_006714126.3 Intron XP_006714189.1

View Full Product Details