Product Details

SNP ID
rs6723477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:181958191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACAGGGAAGTGGAAGTAGCTAA[A/G]CAAGGACTAATTTCAGGTCAACTCC
Phenotype
MIM: 613240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R1C PubMed Links

Gene Details

Gene
PPP1R1C
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080545.2 Intron NP_001074014.1
NM_001261424.1 Intron NP_001248353.1
NM_001261425.1 Intron NP_001248354.1
XM_017003451.1 Intron XP_016858940.1

View Full Product Details