Product Details

SNP ID
rs6751595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:157533006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTAACCATGATATTTCTATGA[G/T]AAAAAATGTCAACAGATATGGTTCT
Phenotype
MIM: 608981
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACVR1C PubMed Links

Gene Details

Gene
ACVR1C
Gene Name
activin A receptor type 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111031.1 2637 UTR 3 NP_001104501.1
NM_001111032.1 2637 UTR 3 NP_001104502.1
NM_001111033.1 2637 UTR 3 NP_001104503.1
NM_145259.2 2637 UTR 3 NP_660302.2

View Full Product Details