Product Details
- SNP ID
-
rs6797951
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:191202643 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGTAAACAGTGAAGGGGGTCGTGA[A/T]ATTCCACATTAACTTCAGATTTAAA
- Phenotype
-
MIM: 610280
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OSTN
PubMed Links
Gene Details
- Gene
- OSTN
- Gene Name
- osteocrin
View Full Product Details