Product Details
- SNP ID
-
rs6770507
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:112926371 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGTTTTTAAATACTGAATTTCTCC[A/T]TATGTTATATGAAAATTAAATGTTT
- Phenotype
-
MIM: 607546
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CD200R1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs375566885] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CD200R1
- Gene Name
- CD200 receptor 1
View Full Product Details