Product Details

SNP ID
rs6747504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70778954 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGGTAAACAGAAATCCTTGCTCA[C/T]TCAGCTTTAGAGTTCAGTCCCTGTT
Phenotype
MIM: 608697
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FIGLA PubMed Links

Gene Details

Gene
FIGLA
Gene Name
folliculogenesis specific bHLH transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004311.3 Intron NP_001004311.2

View Full Product Details