Product Details

SNP ID
rs7967805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:106791060 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTGAGTGAGTGGAGGCCAGAATT[C/T]CTGTGACTTGTACATATTGTTTGGT
Phenotype
MIM: 609147
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RIC8B PubMed Links

Gene Details

Gene
RIC8B
Gene Name
RIC8 guanine nucleotide exchange factor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005268998.3 Intron XP_005269055.1
XM_011538526.2 Intron XP_011536828.1
XM_011538527.2 Intron XP_011536829.1
XM_011538528.2 Intron XP_011536830.1
XM_011538529.2 Intron XP_011536831.1
XM_017019548.1 Intron XP_016875037.1
XM_017019549.1 Intron XP_016875038.1

View Full Product Details