Product Details

SNP ID
rs7668738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:112287558 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGGTTGGGGACCCTTGCCATAGA[A/G]GGAGAAAGAAGACTGAGGATAGATA
Phenotype
MIM: 607347 MIM: 609028
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALPK1 PubMed Links

Gene Details

Gene
ALPK1
Gene Name
alpha kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102406.1 Intron NP_001095876.1
NM_001253884.1 Intron NP_001240813.1
NM_025144.3 Intron NP_079420.3
XM_005263245.4 Intron XP_005263302.1
XM_005263246.4 Intron XP_005263303.1
XM_006714326.3 Intron XP_006714389.1
XM_017008633.1 Intron XP_016864122.1
XM_017008634.1 Intron XP_016864123.1
XM_017008635.1 Intron XP_016864124.1
XM_017008636.1 Intron XP_016864125.1
XM_017008637.1 Intron XP_016864126.1
XM_017008638.1 Intron XP_016864127.1
XM_017008639.1 Intron XP_016864128.1
XM_017008640.1 Intron XP_016864129.1
XM_017008641.1 Intron XP_016864130.1
XM_017008642.1 Intron XP_016864131.1
XM_017008643.1 Intron XP_016864132.1
XM_017008644.1 Intron XP_016864133.1
XM_017008645.1 Intron XP_016864134.1
XM_017008646.1 Intron XP_016864135.1
XM_017008647.1 Intron XP_016864136.1
XM_017008648.1 Intron XP_016864137.1
XM_017008649.1 Intron XP_016864138.1
XM_017008650.1 Intron XP_016864139.1
XM_017008651.1 Intron XP_016864140.1
Gene
TIFA
Gene Name
TRAF interacting protein with forkhead associated domain
There are no transcripts associated with this gene.

View Full Product Details