Product Details

SNP ID
rs6723514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:161316680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCAAATTATGAATGTGATTGAT[A/C]ACCAGATTTTGAAAAGTGGTGTGAT
Phenotype
MIM: 607173
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PSMD14 PubMed Links

Gene Details

Gene
PSMD14
Gene Name
proteasome 26S subunit, non-ATPase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005805.5 Intron NP_005796.1

View Full Product Details