Product Details

SNP ID
rs6658935
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:26242421 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAATCTCATGCCGCTCAGAAACTC[A/G]GATTGTTGCCTGTGTCTTCTATAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP85 PubMed Links

Gene Details

Gene
CEP85
Gene Name
centrosomal protein 85
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281517.2 Intron NP_001268446.1
NM_001281518.2 Intron NP_001268447.1
NM_001319944.1 Intron NP_001306873.1
NM_022778.4 Intron NP_073615.2
XM_017002105.1 Intron XP_016857594.1

View Full Product Details