Product Details

SNP ID
rs6893829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:131960004 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTGAGTCACAGCATGGCGACCTC[C/T]TCCACAGAGTTCTAGATGTGCCATT
Phenotype
MIM: 604443
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSL6 PubMed Links

Gene Details

Gene
ACSL6
Gene Name
acyl-CoA synthetase long-chain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009185.2 Intron NP_001009185.1
NM_001205247.1 Intron NP_001192176.1
NM_001205248.1 Intron NP_001192177.1
NM_001205250.1 Intron NP_001192179.1
NM_001205251.1 Intron NP_001192180.1
NM_015256.3 Intron NP_056071.2

View Full Product Details