Product Details

SNP ID
rs7580839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:196808311 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATTAAGGGATGGGTTAATGCAAA[C/T]AGAAGGGAGTTATTTAGAGCTTTCT
Phenotype
MIM: 604888
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2orf66 PubMed Links

Gene Details

Gene
C2orf66
Gene Name
chromosome 2 open reading frame 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213608.2 Intron NP_998773.1
Gene
GTF3C3
Gene Name
general transcription factor IIIC subunit 3
There are no transcripts associated with this gene.

View Full Product Details