Product Details
- SNP ID
-
rs6718714
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:227174018 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGAATAGATTTTGTCTGGTTATTT[A/C]CCATAAGAAATAAATACTTGTCTAA
- Phenotype
-
MIM: 120070
MIM: 120131
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
COL4A3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78373165] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COL4A3
- Gene Name
- collagen type IV alpha 3 chain
- Gene
- COL4A4
- Gene Name
- collagen type IV alpha 4 chain
There are no transcripts associated with this gene.
View Full Product Details