Product Details
- SNP ID
-
rs7123070
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:33705055 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTATGTACTTGCAGAGTAAGAATT[A/T]AAAAAAATAAACCTGAGAGGTAGTT
- Phenotype
-
MIM: 107271
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C11orf91
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113743057] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C11orf91
- Gene Name
- chromosome 11 open reading frame 91
There are no transcripts associated with this gene.
- Gene
- CD59
- Gene Name
- CD59 molecule
View Full Product Details