Product Details

SNP ID
rs7095770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:104339650 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAGACTTTTTGGCTGAGTCTGAA[A/G]GATAAACAGGCAATGAGAAAATGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP58 PubMed Links

Gene Details

Gene
CFAP58
Gene Name
cilia and flagella associated protein 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008723.1 527 Intron NP_001008723.1
XM_011539377.2 527 UTR 5 XP_011537679.1
XM_017015787.1 527 Intron XP_016871276.1
XM_017015788.1 527 Intron XP_016871277.1
XM_017015789.1 527 Intron XP_016871278.1
XM_017015790.1 527 Intron XP_016871279.1
Gene
ITPRIP
Gene Name
inositol 1,4,5-trisphosphate receptor interacting protein
There are no transcripts associated with this gene.

View Full Product Details