Product Details

SNP ID
rs7923385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97357340 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCAGCACATGAACTAGGCAGACC[A/T]CACGTGGTTCCAGCTTCCACACGGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RRP12 PubMed Links

Gene Details

Gene
RRP12
Gene Name
ribosomal RNA processing 12 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145114.1 Intron NP_001138586.1
NM_001284337.1 Intron NP_001271266.1
NM_015179.3 Intron NP_055994.2
XM_011539555.1 Intron XP_011537857.1
XM_011539556.2 Intron XP_011537858.1

View Full Product Details