Product Details

SNP ID
rs6472251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66125699 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAAATAAATATTGAAAGGACTAAG[A/G]TAATTCTCTAAAGATCCCTGATATG
Phenotype
MIM: 606469
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM55 PubMed Links

Gene Details

Gene
TRIM55
Gene Name
tripartite motif containing 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033058.2 Intron NP_149047.2
NM_184085.1 Intron NP_908973.1
NM_184086.1 Intron NP_908974.1
NM_184087.1 Intron NP_908975.1
XM_005251316.4 Intron XP_005251373.1
XM_011517613.2 Intron XP_011515915.1
XM_011517614.2 Intron XP_011515916.1
XM_011517615.2 Intron XP_011515917.1
XM_011517616.2 Intron XP_011515918.1
XM_011517617.2 Intron XP_011515919.1
XM_017013908.1 Intron XP_016869397.1
XM_017013909.1 Intron XP_016869398.1

View Full Product Details