Product Details

SNP ID
rs7050116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:14880990 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCTATTCCCTATAAGATATATT[C/G]ATTGTTTAGCACAACCTAATTCCCC
Phenotype
MIM: 300515
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FANCB PubMed Links

Gene Details

Gene
FANCB
Gene Name
Fanconi anemia complementation group B
There are no transcripts associated with this gene.

Gene
MOSPD2
Gene Name
motile sperm domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177475.1 Intron NP_001170946.1
NM_152581.3 Intron NP_689794.1
XM_005274451.1 Intron XP_005274508.1

View Full Product Details