Product Details

SNP ID
rs7050041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15324612 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGTAAGAAAAAGCAAATGTGTAC[A/G]TGAAACATCAAGTAAGAGTTCAGAC
Phenotype
MIM: 300626 MIM: 311770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB11 PubMed Links

Gene Details

Gene
ASB11
Gene Name
ankyrin repeat and SOCS box containing 11
There are no transcripts associated with this gene.

Gene
PIGA
Gene Name
phosphatidylinositol glycan anchor biosynthesis class A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002641.3 Intron NP_002632.1
NM_020473.3 Intron NP_065206.3
XM_011545539.2 Intron XP_011543841.1
XM_017029581.1 Intron XP_016885070.1

View Full Product Details