Product Details

SNP ID
rs4300488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:25247312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTACCTTTAAATCATATATGAA[A/G]TAATTTACATGCTATACACTTCCAA
Phenotype
MIM: 603561
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTMR6 PubMed Links

Gene Details

Gene
MTMR6
Gene Name
myotubularin related protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004685.3 3370 UTR 3 NP_004676.3
XM_011535306.1 3370 UTR 3 XP_011533608.1
XM_011535307.1 3370 UTR 3 XP_011533609.1
XM_017020845.1 3370 UTR 3 XP_016876334.1
XM_017020846.1 3370 UTR 3 XP_016876335.1

View Full Product Details