Product Details

SNP ID
rs6592081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:82732630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCGAAAGAGGCCAAATGGCCC[C/G]GCCCGTCCTCCCCGCCGGGAGAATC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM181B PubMed Links

Gene Details

Gene
FAM181B
Gene Name
family with sequence similarity 181 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175885.3 1235 Missense Mutation CCG,CGG P367R NP_787081.2

View Full Product Details