Product Details

SNP ID
rs7461382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42281566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCACTGTAGGGAGTAGGGTGAA[A/G]AAGTCATGGCCCTCTTAGCGTAGAC
Phenotype
MIM: 603258
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IKBKB PubMed Links

Gene Details

Gene
IKBKB
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190720.2 Intron NP_001177649.1
NM_001242778.1 Intron NP_001229707.1
NM_001556.2 Intron NP_001547.1
XM_005273490.2 Intron XP_005273547.1
XM_005273491.4 Intron XP_005273548.1
XM_005273492.3 Intron XP_005273549.1
XM_005273493.3 Intron XP_005273550.1
XM_005273494.2 Intron XP_005273551.1
XM_005273495.2 Intron XP_005273552.1
XM_005273496.3 Intron XP_005273553.1
XM_005273498.3 Intron XP_005273555.1
XM_011544517.2 Intron XP_011542819.1
XM_011544518.2 Intron XP_011542820.1
XM_011544519.2 Intron XP_011542821.1
XM_011544520.2 Intron XP_011542822.1
XM_011544521.2 Intron XP_011542823.1
XM_011544522.2 Intron XP_011542824.1
XM_017013396.1 Intron XP_016868885.1

View Full Product Details