Product Details

SNP ID
rs6923405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29425703 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCAAAATGTTGATGTTGAAGCTG[C/G]ATGGTGGCTACATGGAGTACATGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR11A1 PubMed Links

Gene Details

Gene
OR11A1
Gene Name
olfactory receptor family 11 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013937.3 2030 UTR 3 NP_039225.1
Gene
OR5V1
Gene Name
olfactory receptor family 5 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030876.5 2030 Intron NP_110503.3
XM_011514929.2 2030 Intron XP_011513231.1
XM_011514930.2 2030 Intron XP_011513232.1

View Full Product Details