Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001314017.1 | 1201 | Intron | NP_001300946.1 | ||
NM_001314018.1 | 1201 | UTR 3 | NP_001300947.1 | ||
NM_001314019.1 | 1201 | UTR 3 | NP_001300948.1 | ||
NM_001314020.1 | 1201 | UTR 3 | NP_001300949.1 | ||
NM_020939.1 | 1201 | UTR 3 | NP_065990.1 | ||
XM_005249247.2 | 1201 | UTR 3 | XP_005249304.1 | ||
XM_011514768.1 | 1201 | Intron | XP_011513070.1 | ||
XM_011514769.1 | 1201 | Intron | XP_011513071.1 | ||
XM_011514770.1 | 1201 | Intron | XP_011513072.1 | ||
XM_011514771.2 | 1201 | Intron | XP_011513073.1 | ||
XM_011514772.1 | 1201 | Intron | XP_011513074.1 | ||
XM_011514773.2 | 1201 | Intron | XP_011513075.1 | ||
XM_011514775.2 | 1201 | Intron | XP_011513077.1 | ||
XM_017011139.1 | 1201 | Intron | XP_016866628.1 |