Product Details

SNP ID
rs6904582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39335979 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAGAGGCTGATGGTGAAAGATGT[A/G]GTCTGGGAGATCAGGATTGGCCTTC
Phenotype
MIM: 613919
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF6 PubMed Links

Gene Details

Gene
KIF6
Gene Name
kinesin family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289020.1 1566 UTR 3 NP_001275949.1
NM_001289021.1 1566 UTR 3 NP_001275950.1
NM_001289024.1 1566 UTR 3 NP_001275953.1
NM_145027.4 1566 UTR 3 NP_659464.3
XM_005248904.4 1566 Intron XP_005248961.1
XM_011514357.2 1566 Intron XP_011512659.1
XM_011514358.2 1566 Intron XP_011512660.1
XM_011514359.2 1566 Intron XP_011512661.1
XM_011514361.2 1566 Intron XP_011512663.1
XM_017010427.1 1566 Intron XP_016865916.1
XM_017010428.1 1566 Intron XP_016865917.1

View Full Product Details