Product Details

SNP ID
rs6912670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37214947 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCGCAGCTTCCAGAGCAGCCACTA[A/C]CCATGGGGTACTCACCTGAATTGTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM217 PubMed Links

Gene Details

Gene
TMEM217
Gene Name
transmembrane protein 217
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162900.1 Intron NP_001156372.1
NM_001286401.1 Intron NP_001273330.1
NM_145316.3 Intron NP_660359.2
XM_011514362.1 Intron XP_011512664.1
XM_011514363.2 Intron XP_011512665.1
XM_011514364.2 Intron XP_011512666.1
XM_011514365.2 Intron XP_011512667.1
XM_011514366.2 Intron XP_011512668.1
XM_011514367.2 Intron XP_011512669.1
XM_011514369.2 Intron XP_011512671.1
XM_011514370.2 Intron XP_011512672.1

View Full Product Details