Product Details

SNP ID
rs7739288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52742349 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTGGTGAAATTATTTATACAGTT[A/T]CAACCAGTGCAAACTTTTTCTTTTG
Phenotype
MIM: 138360
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GSTA2 PubMed Links

Gene Details

Gene
GSTA2
Gene Name
glutathione S-transferase alpha 2
There are no transcripts associated with this gene.

Gene
GSTA7P
Gene Name
glutathione S-transferase alpha 7, pseudogene
There are no transcripts associated with this gene.

View Full Product Details