Product Details

SNP ID
rs8037482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48196099 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTACTTTCTATCATTTGCACCCCT[A/G]TTCAATTTAAGACATTTGCTTTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTXN2 PubMed Links

Gene Details

Gene
CTXN2
Gene Name
cortexin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145668.1 Intron NP_001139140.1
XM_005254378.3 Intron XP_005254435.1
XM_006720504.2 Intron XP_006720567.1
XM_011521553.1 Intron XP_011519855.1
XM_017022178.1 Intron XP_016877667.1

View Full Product Details