Product Details

SNP ID
rs6491313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:96883588 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAGAGAAAAAACACTAGAAATCA[C/T]TGGAGTGAGGCAGCTGGGACAGGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSP90AB6P PubMed Links

Gene Details

Gene
HSP90AB6P
Gene Name
heat shock protein 90 alpha family class B member 6, pseudogene
There are no transcripts associated with this gene.

View Full Product Details