Product Details

SNP ID
rs3864919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:16189366 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGTCATACGCTTTCAATGGCTG[A/C]ACCATTGTAGAAAGCTGTAAGGTAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC15A5 PubMed Links

Gene Details

Gene
SLC15A5
Gene Name
solute carrier family 15 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170798.1 2042 UTR 3 NP_001164269.1

View Full Product Details