Product Details

SNP ID
rs4355029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:62327139 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGGTCTCTGACCAGCCCTTCGGA[C/G]AATAGGTGAGAGCCTGTGTGGGCTG
Phenotype
MIM: 603499
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TNFRSF11A PubMed Links

Gene Details

Gene
TNFRSF11A
Gene Name
TNF receptor superfamily member 11a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270949.1 Intron NP_001257878.1
NM_001270950.1 Intron NP_001257879.1
NM_001270951.1 Intron NP_001257880.1
NM_001278268.1 Intron NP_001265197.1
NM_003839.3 Intron NP_003830.1
XM_011526244.2 Intron XP_011524546.1
XM_011526245.2 Intron XP_011524547.1
XM_017026064.1 Intron XP_016881553.1
XM_017026065.1 Intron XP_016881554.1
XM_017026066.1 Intron XP_016881555.1

View Full Product Details