Product Details

SNP ID
rs6502180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2345989 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCTTGCGGGGGACGGGGAGGGAA[C/G]ACTAGCAAGGTATGGATCAAAAACA
Phenotype
MIM: 611418 MIM: 611214
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SGSM2 PubMed Links

Gene Details

Gene
SGSM2
Gene Name
small G protein signaling modulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098509.1 Intron NP_001091979.1
NM_014853.2 Intron NP_055668.2
XM_011524101.2 Intron XP_011522403.1
XM_011524102.2 Intron XP_011522404.1
XM_011524103.2 Intron XP_011522405.1
XM_011524104.2 Intron XP_011522406.1
XM_011524105.2 Intron XP_011522407.1
XM_011524106.1 Intron XP_011522408.1
XM_011524107.2 Intron XP_011522409.1
XM_011524108.1 Intron XP_011522410.1
XM_011524109.2 Intron XP_011522411.1
XM_011524110.2 Intron XP_011522412.1
XM_017025474.1 Intron XP_016880963.1
XM_017025475.1 Intron XP_016880964.1
XM_017025476.1 Intron XP_016880965.1
Gene
TSR1
Gene Name
TSR1, ribosome maturation factor
There are no transcripts associated with this gene.

View Full Product Details