Product Details

SNP ID
rs6428896
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:116381170 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCAATGCCCAGATGAGTGGGAAG[A/G]TTACTGTAGTTGATTGAGGAAATGC
Phenotype
MIM: 182310
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP1A1 PubMed Links

Gene Details

Gene
ATP1A1
Gene Name
ATPase Na+/K+ transporting subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000701.7 3217 Intron NP_000692.2
NM_001160233.1 3217 Intron NP_001153705.1
NM_001160234.1 3217 Intron NP_001153706.1
XM_017001360.1 3217 UTR 5 XP_016856849.1
XM_017001361.1 3217 Intron XP_016856850.1

View Full Product Details